missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isoamyl Isobutyrate, TCI America™

$32.08 - $377.38

Chemical Identifiers

CAS 2050-01-3
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00053719
InChI Key VFTGLSWXJMRZNB-UHFFFAOYSA-N
Synonym isoamyl isobutyrate, isopentyl isobutyrate, isobutyric acid isoamyl ester, propanoic acid, 2-methyl-, 3-methylbutyl ester, isobutyric acid, isopentyl ester, 3-methylbutyl isobutyrate, isoamyl 2-methylpropanoate, isopentyl 2-methylpropanoate, isoamyl isobutanoate, isopentyl isobutanoate
PubChem CID 519786
ChEBI CHEBI:87537
IUPAC Name 3-methylbutyl 2-methylpropanoate
SMILES CC(C)CCOC(=O)C(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I0676250G View Documents TCI America
I0676250G
250g Each for $195.56
 
I067625G View Documents TCI America
I067625G
25g Each for $32.08
 
I0676500G View Documents TCI America
I0676500G
500g Each for $377.38
 
Specifications

Chemical Identifiers

2050-01-3
158.24
VFTGLSWXJMRZNB-UHFFFAOYSA-N
519786
3-methylbutyl 2-methylpropanoate
C9H18O2
MFCD00053719
isoamyl isobutyrate, isopentyl isobutyrate, isobutyric acid isoamyl ester, propanoic acid, 2-methyl-, 3-methylbutyl ester, isobutyric acid, isopentyl ester, 3-methylbutyl isobutyrate, isoamyl 2-methylpropanoate, isopentyl 2-methylpropanoate, isoamyl isobutanoate, isopentyl isobutanoate
CHEBI:87537
CC(C)CCOC(=O)C(C)C

Specifications

169°C
250g
C9H18O2
2620
VFTGLSWXJMRZNB-UHFFFAOYSA-N
3-methylbutyl 2-methylpropanoate
519786
158.24
≥98.0% (GC)
Colorless
2050-01-3
MFCD00053719
isoamyl isobutyrate, isopentyl isobutyrate, isobutyric acid isoamyl ester, propanoic acid, 2-methyl-, 3-methylbutyl ester, isobutyric acid, isopentyl ester, 3-methylbutyl isobutyrate, isoamyl 2-methylpropanoate, isopentyl 2-methylpropanoate, isoamyl isobutanoate, isopentyl isobutanoate
CC(C)CCOC(=O)C(C)C
158.24
CHEBI:87537
Clear Liquid at 20°C
Isoamyl Isobutyrate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0779

TSCA : Yes