Please login to your online account to display your discounted pricing

Isoamyl Alcohol (Clear, Colorless/Certified), Fisher Chemical

$202.31 - $708.77

Chemical Identifiers

Chemical Name or Material Isoamyl Alcohol
CAS 123-51-3
Purity Grade Certified Grade
Physical Form Liquid
Molecular Formula C5H12O
Linear Formula (CH3)2CHCH2CH2OH
Formula Weight 88.15
MDL Number MFCD00002934
Synonym 3-Methyl-1-butanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

A393-500

  SDS
fisher scientific company
BPA393500
Glass Bottle 500mL Each for $202.31
Case of 6 Each for $1,213.86

A393-4

  Accessories SDS
fisher scientific company
BPA3934
Glass Bottle 4L Each for $708.77
Case of 4 Each for $2,835.08
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 88.15
Boiling Point 130°C
Melting Point −117°C
Quantity 500mL
Density 3.04 (Air = 1.0)
UN Number 1105
Flash Point 43°C
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Molecular Formula C5H12O
Vapor Pressure 4 hPa at 20°C
Color Clear
pH 6.5
Packaging Glass Bottle
Acidity 0.01% max.
Cleanup Symbol Solvent
MDL Number MFCD00002934
ChemAlert Storage Symbol Red
Synonym 3-Methyl-1-butanol
Residue after Evaporation 0.003% max.
Additional Information Vapor Pressure: 4hPa at 20°C
Water 0.5% max.
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Irritating to eyes and respiratory system. Aspiration hazard if swallowed - can enter lungs and cause damage. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:2
Instability:0
Chemical Name or Material Isoamyl Alcohol
Carbonyl Compounds 0.1% max.
Linear Formula (CH3)2CHCH2CH2OH
CAS 123-51-3
Health Hazard 2 WARNING!

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Emergency Overview
Flammable liquid and vapor. Irritating to eyes and respiratory system. Aspiration hazard if swallowed - can enter lungs and cause damage. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:2
Instability:0

WARNING!