Please login to your online account to display your discounted pricing

iso-Propyl Ether, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 1L
Packaging Aluminum Bottle

Manufacturer: avantor performance  924322

 View more versions of this product

Catalog No. 02003288

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 102.18
Chemical Name or Material iso-Propyl Ether
Molecular Formula (CH3)2CHOCH(CH3)2
CAS 108-20-3
Packaging Aluminum Bottle
Specific Gravity 1L = 0.78kg
Flash Point −28.00°C
Quantity 1L

Specifications

Chemical Name or Material iso-Propyl Ether
CAS 108-20-3
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula (CH3)2CHOCH(CH3)2
Formula Weight 102.18