Please login to your online account to display your discounted pricing

iso-Propyl Ether, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$432.67 - $756.85

Chemical Identifiers

Chemical Name or Material iso-Propyl Ether
CAS 108-20-3
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula (CH3)2CHOCH(CH3)2
Formula Weight 102.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003288

 
avantor performance
924322
Aluminum Bottle 1L Each for $432.67

02003289

 
avantor performance
924303
Aluminum Bottle 4L Each for $756.85
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 102.18
Chemical Name or Material iso-Propyl Ether
Molecular Formula (CH3)2CHOCH(CH3)2
CAS 108-20-3
Packaging Aluminum Bottle
Specific Gravity 1L = 0.78kg
Flash Point −28.00°C
Quantity 1L