Please login to your online account to display your discounted pricing

iso-Propyl Ether, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 4L
Packaging Aluminum Bottle

Manufacturer: avantor performance  924303

 View more versions of this product

Catalog No. 02003289

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 102.18
Purity Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material iso-Propyl Ether
CAS 108-20-3
Molecular Formula (CH3)2CHOCH(CH3)2
Specific Gravity 1L = 0.78kg
Flash Point −28.00°C
Packaging Aluminum Bottle
Quantity 4L

Specifications

Chemical Name or Material iso-Propyl Ether
CAS 108-20-3
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula (CH3)2CHOCH(CH3)2
Formula Weight 102.18