Please login to your online account to display your discounted pricing

iso-Propyl Alcohol ACS, EMD Millipore


Quantity 4L
Packaging Glass Bottle

Manufacturer: emd millipore corporation  PX18355*

 View more versions of this product

Catalog No. MPX18355

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 60.1
Purity Grade ACS Reagent Grade
Quantity 4L
Linear Formula CH3CHOHCH3
Packaging Glass Bottle
Chemical Name or Material iso-Propyl Alcohol

Specifications

Chemical Name or Material iso-Propyl Alcohol
Purity Grade ACS Reagent Grade
Linear Formula CH3CHOHCH3
Formula Weight 60.1