Please login to your online account to display your discounted pricing

iso-PPADS tetrasodium salt, Tocris


P2X antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS068310

 View more versions of this product

Catalog No. 068310

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 599.3
Assay Percent Range >98%
Molecular Formula C14H10N3Na4O12PS2
Chemical Name or Material iso-PPADS tetrasodium salt
CAS 207572-67-6
Synonym Pyridoxalphosphate-6-azophenyl-2', 5'-disulfonic acid tetrasodium salt
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material iso-PPADS tetrasodium salt
Assay Percent Range >98%
Molecular Formula C14H10N3Na4O12PS2
Formula Weight 599.3
Synonym Pyridoxalphosphate-6-azophenyl-2', 5'-disulfonic acid tetrasodium salt
Accessories

Desiccate at -20°C