Please login to your online account to display your discounted pricing

iso-PPADS tetrasodium salt, Tocris

P2X antagonist

$116.01 - $488.83

Chemical Identifiers

Chemical Name or Material iso-PPADS tetrasodium salt
Assay Percent Range >98%
Molecular Formula C14H10N3Na4O12PS2
Formula Weight 599.3
Synonym Pyridoxalphosphate-6-azophenyl-2', 5'-disulfonic acid tetrasodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

068310

 
r & d systems
RDS068310
10mg Each for $116.01

068350

 
r & d systems
RDS068350
50mg Each for $488.83
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 599.3
Assay Percent Range >98%
Molecular Formula C14H10N3Na4O12PS2
Chemical Name or Material iso-PPADS tetrasodium salt
CAS 207572-67-6
Synonym Pyridoxalphosphate-6-azophenyl-2', 5'-disulfonic acid tetrasodium salt
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C