missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ishikawa's Reagent, 90+%

$106.72 - $359.31

Chemical Identifiers

CAS 309-88-6
Molecular Formula C7H11F6N
Molecular Weight (g/mol) 223.162
MDL Number MFCD00054683
InChI Key BNTFCVMJHBNJAR-UHFFFAOYSA-N
Synonym n,n-diethyl-1,1,2,3,3,3-hexafluoropropylamine, n,n-diethyl 1,1,2,3,3,3-hexafluoropropylamine, diethyl 1,1,2,3,3,3-hexafluoropropyl amine, 1,1,2,3,3,3-hexafluoro-1-diethylaminopropane, n,n-diethyl-1,1,2,3,3,3-hexafluoro-1-propanamine, 1-propanamine, n,n-diethyl-1,1,2,3,3,3-hexafluoro, n,n-diethyl-1,1,2,3,3,3-hexafluoropropyl amine, ishikawa s reagent, acmc-20ak9c, ishikawa's reagant
PubChem CID 136149
IUPAC Name N,N-diethyl-1,1,2,3,3,3-hexafluoropropan-1-amine
SMILES CCN(CC)C(C(C(F)(F)F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1673814 View Documents Alfa Aesar™
L1673814
25g Each for $106.72
 
AAL1673822 View Documents Alfa Aesar™
L1673822
100g Each for $359.31
 
Specifications

Chemical Identifiers

309-88-6
223.162
BNTFCVMJHBNJAR-UHFFFAOYSA-N
136149
CCN(CC)C(C(C(F)(F)F)F)(F)F
C7H11F6N
MFCD00054683
n,n-diethyl-1,1,2,3,3,3-hexafluoropropylamine, n,n-diethyl 1,1,2,3,3,3-hexafluoropropylamine, diethyl 1,1,2,3,3,3-hexafluoropropyl amine, 1,1,2,3,3,3-hexafluoro-1-diethylaminopropane, n,n-diethyl-1,1,2,3,3,3-hexafluoro-1-propanamine, 1-propanamine, n,n-diethyl-1,1,2,3,3,3-hexafluoro, n,n-diethyl-1,1,2,3,3,3-hexafluoropropyl amine, ishikawa s reagent, acmc-20ak9c, ishikawa's reagant
N,N-diethyl-1,1,2,3,3,3-hexafluoropropan-1-amine

Specifications

14°C (57°F)
25g
C7H11F6N
MFCD00054683
n,n-diethyl-1,1,2,3,3,3-hexafluoropropylamine, n,n-diethyl 1,1,2,3,3,3-hexafluoropropylamine, diethyl 1,1,2,3,3,3-hexafluoropropyl amine, 1,1,2,3,3,3-hexafluoro-1-diethylaminopropane, n,n-diethyl-1,1,2,3,3,3-hexafluoro-1-propanamine, 1-propanamine, n,n-diethyl-1,1,2,3,3,3-hexafluoro, n,n-diethyl-1,1,2,3,3,3-hexafluoropropyl amine, ishikawa s reagent, acmc-20ak9c, ishikawa's reagant
CCN(CC)C(C(C(F)(F)F)F)(F)F
223.162
223.16
Ishikawa's Reagent
56°C to 57°C (58mmHg)
309-88-6
CF3CFHCF2N(CH2CH3)2
UN2924
BNTFCVMJHBNJAR-UHFFFAOYSA-N
N,N-diethyl-1,1,2,3,3,3-hexafluoropropan-1-amine
136149
≥90%
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.