missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ISG15 Armenian Hamster anti-Mouse, Clone: 3C2, eBioscience™

Armenian Hamster Monoclonal Antibody

$183.48 - $368.28

Specifications

Antigen ISG15
Clone 3C2
Host Species Armenian Hamster
Gene Alias ISG-15
Applications Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013219 Life Technologies
LS14585780
25μg Each for $183.48
5013220 Life Technologies
LS14585782
100μg Each for $368.28
Specifications

Description

This 3C2 monoclonal antibody reacts with mouse interferon stimulated gene-15 (ISG-15), a 15kDa ubiquitin-like protein (UBL) and cytokine. ISG-15 is translated as a precursor that is subsequently activated by an E1 enzyme, conjugated by an E2 enzyme via the C-terminal LRLRGG motif and ligated by an E3 ligase. The activity of ISG-15 is dependent on its modification of target proteins at specific lysine residues. Several viruses utilize ISG-15 and ubiquitin deconjugating proteases, while influenza B protein NS1 has been shown to specifically bind ISG-15 thereby blocking its conjugation.

In addition to its role as a protein modification, ISG-15 is secreted by monocytes and lymphocytes in response to IFNα, β, or γ, as well as influenza B virus. In this role as cytokine, it has been shown to induce secretion of IFNγ and support NK cell proliferation and cytolytic activity.
Description & Specifications

Specifications

ISG15
Armenian Hamster
Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
3C2
ISG-15
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer