missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Isethionic acid, sodium salt, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 205005000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Isethionic acid, sodium salt | |
1562-00-1 | |
98% | |
C2H5NaO4S | |
MFCD00007534 | |
04, III, 42 | |
sodium 2-hydroxyethanesulfonate, sodium isethionate, isethionic acid sodium salt, 2-hydroxyethanesulfonic acid sodium salt, sodium hydroxyethylsulfonate, ethanesulfonic acid, 2-hydroxy-, monosodium salt, sodium 2-hydroxyethylsulfonate, sodium 2-hydroxyethanesulphonate, sodium beta-hydroxyethanesulfonate, sodium 1-hydroxy-2-ethanesulfonate | |
LADXKQRVAFSPTR-UHFFFAOYSA-M | |
sodium;2-hydroxyethanesulfonate | |
517063 | |
98% |
98% | |
Authentic | |
Plastic bottle | |
HOCH2CH2SO3Na | |
500g | |
14, 5107 | |
Solubility in water: soluble in water | |
C(CS(=O)(=O)[O-])O.[Na+] | |
148.11 | |
148.11 |
Chemical Identifiers
1562-00-1 | |
148.11 | |
LADXKQRVAFSPTR-UHFFFAOYSA-M | |
517063 | |
C(CS(=O)(=O)[O-])O.[Na+] |
C2H5NaO4S | |
MFCD00007534 | |
sodium 2-hydroxyethanesulfonate, sodium isethionate, isethionic acid sodium salt, 2-hydroxyethanesulfonic acid sodium salt, sodium hydroxyethylsulfonate, ethanesulfonic acid, 2-hydroxy-, monosodium salt, sodium 2-hydroxyethylsulfonate, sodium 2-hydroxyethanesulphonate, sodium beta-hydroxyethanesulfonate, sodium 1-hydroxy-2-ethanesulfonate | |
sodium;2-hydroxyethanesulfonate |
Safety and Handling
HYGROSCOPIC
EINECSNumber : 216-343-6
TSCA : TSCA
RUO â Research Use Only