Learn More
Isethionic acid sodium salt, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A120540B
Description
Isethionic acid sodium salt is an amphoteric detergent used in detergent bar soaps. It makes a dense lather in addition to the lather made by the soap. It is mild on the skin, and non-drying. It works equally well in soft or hard water. It is also an anti-static agent in shampoos.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsethionic acid sodium salt is an amphoteric detergent used in detergent bar soaps. It makes a dense lather in addition to the lather made by the soap. It is mild on the skin, and non-drying. It works equally well in soft or hard water. It is also an anti-static agent in shampoos.
Solubility
Soluble in water.
Notes
Store in cool place. Keep away from oxidizing agents.
Specifications
Isethionic acid sodium salt | |
191°C to 194°C | |
MFCD00007534 | |
3633992 | |
Soluble in water. | |
C(CS(=O)(=O)[O-])O.[Na+] | |
148.108 | |
148.11 |
1562-00-1 | |
C2H5NaO4S | |
1000 g | |
sodium 2-hydroxyethanesulfonate, sodium isethionate, isethionic acid sodium salt, 2-hydroxyethanesulfonic acid sodium salt, sodium hydroxyethylsulfonate, ethanesulfonic acid, 2-hydroxy-, monosodium salt, sodium 2-hydroxyethylsulfonate, sodium 2-hydroxyethanesulphonate, sodium beta-hydroxyethanesulfonate, sodium 1-hydroxy-2-ethanesulfonate | |
LADXKQRVAFSPTR-UHFFFAOYSA-M | |
sodium;2-hydroxyethanesulfonate | |
517063 | |
98% |
Chemical Identifiers
1562-00-1 | |
148.108 | |
LADXKQRVAFSPTR-UHFFFAOYSA-M | |
517063 | |
C(CS(=O)(=O)[O-])O.[Na+] |
C2H5NaO4S | |
MFCD00007534 | |
sodium 2-hydroxyethanesulfonate, sodium isethionate, isethionic acid sodium salt, 2-hydroxyethanesulfonic acid sodium salt, sodium hydroxyethylsulfonate, ethanesulfonic acid, 2-hydroxy-, monosodium salt, sodium 2-hydroxyethylsulfonate, sodium 2-hydroxyethanesulphonate, sodium beta-hydroxyethanesulfonate, sodium 1-hydroxy-2-ethanesulfonate | |
sodium;2-hydroxyethanesulfonate |
Safety and Handling
EINECSNumber : 216-343-6
RTECSNumber : KI7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only