Please login to your online account to display your discounted pricing

Isethionic acid, potassium salt, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  412630250

 View more versions of this product

Catalog No. AC412630250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 164.22
Physical Form Crystalline Powder
Quantity 25g
Color White
Packaging Glass bottle
Molecular Formula C2H5KO4S
Synonym 2-Hydroxyethanesulfonic acid potassium salt
Chemical Name or Material Isethionic acid, potassium salt
Assay Percent Range 98%
Infrared Spectrum Authentic
Appearance of Solution (2% aq. soln.) clear, colorless
Additional Information Remark: may contain small amounts of sodium salt
Linear Formula HOCH2CH2SO3K
CAS 1561-99-5

Specifications

Chemical Name or Material Isethionic acid, potassium salt
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C2H5KO4S
Linear Formula HOCH2CH2SO3K
Formula Weight 164.22
Synonym 2-Hydroxyethanesulfonic acid potassium salt