Please login to your online account to display your discounted pricing

Isethionic acid, potassium salt, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Isethionic acid, potassium salt
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C2H5KO4S
Linear Formula HOCH2CH2SO3K
Formula Weight 164.22
Synonym 2-Hydroxyethanesulfonic acid potassium salt
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC412630250

  SDS
fisher scientific acros
412630250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC412631000

  SDS
thermo fisher scientific
AC412631000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 164.22
Physical Form Crystalline Powder
Quantity 25g
Color White
Packaging Glass bottle
Molecular Formula C2H5KO4S
Synonym 2-Hydroxyethanesulfonic acid potassium salt
Chemical Name or Material Isethionic acid, potassium salt
Assay Percent Range 98%
Infrared Spectrum Authentic
Appearance of Solution (2% aq. soln.) clear, colorless
Additional Information Remark: may contain small amounts of sodium salt
Linear Formula HOCH2CH2SO3K
CAS 1561-99-5