Please login to your online account to display your discounted pricing

Isethionic acid, potassium salt, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC412631000

 View more versions of this product

Catalog No. AC412631000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Isethionic acid, potassium salt
Quantity 100g
CAS 1561-99-5
Additional Information Remark: may contain small amounts of sodium salt
Appearance of Solution (2% aq. soln.) clear, colorless
Physical Form Crystalline Powder
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C2H5KO4S
Linear Formula HOCH2CH2SO3K
Formula Weight 164.22
Synonym 2-Hydroxyethanesulfonic acid potassium salt
Infrared Spectrum Authentic
Packaging Glass bottle
Color White

Specifications

Chemical Name or Material Isethionic acid, potassium salt
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C2H5KO4S
Linear Formula HOCH2CH2SO3K
Formula Weight 164.22
Synonym 2-Hydroxyethanesulfonic acid potassium salt