Please login to your online account to display your discounted pricing

Alfa Aesar™ Isethionic acid ammonium salt, 99%

$26.81 - $66.45

Chemical Identifiers

Chemical Name or Material Isethionic acid ammonium salt
CAS 57267-78-4
Molecular Formula HO(CH2)2SO3NH4
Formula Weight 143.16
MDL Number MFCD00012847
Beilstein 3722656
Synonym Ammonium isethionate; 2-Hydroxyethanesulfonic acid ammonium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1435303

  SDS
alfa aesar™
L1435303
1g Each for $26.81

AAL1435306

  SDS
alfa aesar™
L1435306
5g Each for $66.45
Description & Specifications

Specifications

Formula Weight 143.16
MDL Number MFCD00012847
Assay 99%
CAS 57267-78-4
Molecular Formula HO(CH2)2SO3NH4
Beilstein 3722656
Synonym Ammonium isethionate; 2-Hydroxyethanesulfonic acid ammonium salt
TSCA TSCA
RTECS Number KI7750000
Quantity 1g
Melting Point 138° to 139°C
Chemical Name or Material Isethionic acid ammonium salt