missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Isatoic Anhydride, TCI America™

$76.39 - $736.41

Chemical Identifiers

CAS 118-48-9
Molecular Formula C8H5NO3
Molecular Weight (g/mol) 163.132
MDL Number MFCD00006700
InChI Key TXJUTRJFNRYTHH-UHFFFAOYSA-N
Synonym isatoic anhydride, 2h-3,1-benzoxazine-2,4 1h-dione, 1h-benzo d 1,3 oxazine-2,4-dione, isatoic acid anhydride, 4h-3,1-benzoxazine-2,4 1h-dione, isato acid anhydride, n-carboxyanthranilic anhydride, unii-r8tfa74y4u, 3,1-benzoxazine-2,4 1h-dione, anthranilic acid, n-carboxy-, cyclic anhydride
PubChem CID 8359
IUPAC Name 1H-3,1-benzoxazine-2,4-dione
SMILES C1=CC=C2C(=C1)C(=O)OC(=O)N2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I008125G View Documents TCI America
I008125G
25g Each for $76.39
 
I0081500G View Documents TCI America
I0081500G
500g Each for $736.41
 
Specifications

Chemical Identifiers

118-48-9
163.132
TXJUTRJFNRYTHH-UHFFFAOYSA-N
8359
C1=CC=C2C(=C1)C(=O)OC(=O)N2
C8H5NO3
MFCD00006700
isatoic anhydride, 2h-3,1-benzoxazine-2,4 1h-dione, 1h-benzo d 1,3 oxazine-2,4-dione, isatoic acid anhydride, 4h-3,1-benzoxazine-2,4 1h-dione, isato acid anhydride, n-carboxyanthranilic anhydride, unii-r8tfa74y4u, 3,1-benzoxazine-2,4 1h-dione, anthranilic acid, n-carboxy-, cyclic anhydride
1H-3,1-benzoxazine-2,4-dione

Specifications

White
118-48-9
MFCD00006700
TXJUTRJFNRYTHH-UHFFFAOYSA-N
1H-3,1-benzoxazine-2,4-dione
8359
Crystal-Powder at 20°C
Isatoic Anhydride
25g
C8H5NO3
isatoic anhydride, 2h-3,1-benzoxazine-2,4 1h-dione, 1h-benzo d 1,3 oxazine-2,4-dione, isatoic acid anhydride, 4h-3,1-benzoxazine-2,4 1h-dione, isato acid anhydride, n-carboxyanthranilic anhydride, unii-r8tfa74y4u, 3,1-benzoxazine-2,4 1h-dione, anthranilic acid, n-carboxy-, cyclic anhydride
C1=CC=C2C(=C1)C(=O)OC(=O)N2
163.132
163.13
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0856

RTECSNumber : DM3100000

TSCA : Yes