Please login to your online account to display your discounted pricing

Alfa Aesar™ Isatin-3-oxime, 97%

$58.81 - $632.65

Chemical Identifiers

Chemical Name or Material Isatin-3-oxime
CAS 607-28-3
Molecular Formula C8H6N2O2
Formula Weight 162.15
MDL Number MFCD00014568
Beilstein 136198
Synonym beta-Isatoxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2273206

 
alfa aesar™
B2273206
5g Each for $58.81

AAB2273214

 
alfa aesar™
B2273214
25g Each for $190.86

AAB2273222

 
alfa aesar™
B2273222
100g Each for $632.65
Description & Specifications

Specifications

Formula Weight 162.15
Chemical Name or Material Isatin-3-oxime
Assay 97%
Molecular Formula C8H6N2O2
MDL Number MFCD00014568
CAS 607-28-3
Beilstein 136198
Synonym beta-Isatoxime
Melting Point ≈228°C (dec.)
RTECS Number NL7980000
Quantity 5g