Please login to your online account to display your discounted pricing

Iron yttrium oxide, 99.9% (REO), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Iron yttrium oxide
CAS 12063-56-8
Physical Form Powder
Assay Percent Range 99.9% (REO)
Molecular Formula Y3Fe5O12
Formula Weight 737.94
MDL Number MFCD00210618
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3948814

 
alfa aesar
3948814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3948822

 
alfa aesar
3948822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 737.94
Quantity 25g
Color Green
Chemical Name or Material Iron yttrium oxide
EINECS Number 235-053-0
Odor Odorless
Physical Form Powder
Assay Percent Range 99.9% (REO)
Molecular Formula Y3Fe5O12
Appearance Green powder
MDL Number MFCD00210618
CAS 12063-56-8
Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.