Please login to your online account to display your discounted pricing

American Educational Products Iron Wire Gauze

Economical, general-purpose wire gauze — a laboratory staple.

Manufacturer: american educational products  P35076

 View all options for this product

Catalog No. S50151


Description & Specifications


Quantity 12 Pack
Length (English) 5 in.
Length (Metric) 13cm
Width (English) 5 in.
Material Steel
Product Type Wire Gauze
Width (Metric) 13cm
For Use With (Application) Provide good support for flame heating
Dimensions (L x W) 5 x 5 in. (13 x 13cm)

  • Good support for flame heating
  • Fairly durable, inexpensive solution to many classroom needs
  • 16 mesh, no. 25 B&S gauge