Please login to your online account to display your discounted pricing
American Educational Products Iron Wire Gauze
Economical, general-purpose wire gauze — a laboratory staple.
$16.55 - $20.25
Specifications
Product Type Wire Gauze
For Use With (Application) Provide good support for flame heating
Quantity 12 Pack
View More Specs
Products
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity    
S50150
american educational products
7-S50150
4 x 4 in. (10 x 10cm) Pack of 12 Each for $16.55
S50151
american educational products
P35076
5 x 5 in. (13 x 13cm) Pack of 12 Each for $20.25
Description & Specifications
Specifications
Product Type Wire Gauze
For Use With (Application) Provide good support for flame heating
Quantity 12 Pack
  • Good support for flame heating
  • Fairly durable, inexpensive solution to many classroom needs
  • 16 mesh, no. 25 B&S gauge