missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iron wire, greater than 99% (metals basis), Alfa Aesar
Manufacturer: Alfa Aesar 14147H4
Specifications
Iron | |
7439-89-6 | |
7.87g/cm3 at 20°C | |
Fe | |
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m | |
[Fe] | |
55.845 | |
CHEBI:82664 |
100m | |
Gray wire | |
>99% (Metals basis) | |
MFCD00010999 | |
XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
iron | |
23925 | |
Wire, 2mm (0.08 in.) dia. |
Chemical Identifiers
7439-89-6 | |
55.845 | |
XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
23925 | |
iron |
Fe | |
MFCD00010999 | |
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m | |
CHEBI:82664 | |
[Fe] |
Safety and Handling
EINECSNumber : 231-096-4