missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron wire, >99% (metals basis), Alfa Aesar™

Form: approx. 25g/m

Manufacturer:  Alfa Aesar 14246H2

 View more versions of this product

Catalog No. AA14246H2



Specifications

Specifications

Iron
7439-89-6
7.87g/cm3 at 20°C
Fe
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
[Fe]
55.845
CHEBI:82664
Gray
50m
Gray wire
>99% (Metals basis)
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
23925
Wire, 2mm (0.08 in.) dia.

Chemical Identifiers

7439-89-6
55.845
XEEYBQQBJWHFJM-UHFFFAOYSA-N
23925
iron
Fe
MFCD00010999
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
CHEBI:82664
[Fe]
Safety and Handling

Safety and Handling

EINECSNumber : 231-096-4

Documents