missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron wire, 99.998% (metals basis), Alfa Aesar™, Puratronic™

Form: approx. 0.06g/m

Manufacturer:  Thermo Fisher Scientific 10938H2

 View more versions of this product

Catalog No. AA10938H2



Specifications

Specifications

Iron
7439-89-6
99.998% (Metals basis)
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
23925
Wire, 0.1mm (0.004 in.) dia.
50m
Gray wire
Fe
3zhp
[Fe]
55.845
CHEBI:82664
Gray

Chemical Identifiers

7439-89-6
55.845
XEEYBQQBJWHFJM-UHFFFAOYSA-N
23925
iron
Fe
MFCD00010999
3zhp
CHEBI:82664
[Fe]
Safety and Handling

Safety and Handling

EINECSNumber : 231-096-4

Documents