missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron wire,1mm dia., 99.995% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Alfa Aesar™ 10935CB

 View more versions of this product

Catalog No. AA10935CB



Specifications

Specifications

Iron
7439-89-6
99.995% (Metals basis)
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
23925
Wire, 0.25mm (0.01 in.) dia.
250cm
Gray wire
Fe
3zhp
[Fe]
55.845
CHEBI:82664

Chemical Identifiers

7439-89-6
55.845
XEEYBQQBJWHFJM-UHFFFAOYSA-N
23925
iron
Fe
MFCD00010999
3zhp
CHEBI:82664
[Fe]
Safety and Handling

Safety and Handling

EINECSNumber : 231-096-4

Documents