missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron Wire, 2.0mm (0.08 in.) dia., Puratronic™, 99.995% (Metals basis), Alfa Aesar™

Form: Approx.25g/m

Manufacturer:  Thermo Fisher Scientific 10934BU

 View more versions of this product

Catalog No. AA10934BU



Specifications

Specifications

Iron
7439-89-6
99.995% (Metals basis)
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
23925
Wire, 2mm (0.08 in.) dia.
25cm
Gray wire
Fe
3zhp
[Fe]
55.845
CHEBI:82664
Gray

Chemical Identifiers

7439-89-6
55.845
XEEYBQQBJWHFJM-UHFFFAOYSA-N
23925
iron
Fe
MFCD00010999
3zhp
CHEBI:82664
[Fe]
Documents