missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron tris(trifluoromethylsulfonyl)imide

Manufacturer:  Alfa Aesar™ 4718606

 View more versions of this product

Catalog No. AA4718606



Specifications

Specifications

Iron tris(trifluoromethylsulfonyl)imide
C6F18FeN3O12S6
MFCD23380171
Iron tris(trifluoromethanesulfonyl)imide; Iron tris(trifluoromethanesulfonimide)
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Fe+3]
896.251
896.25
Hygroscopic
5g
207861-59-4
[(CF3SO2)2N]3Fe
UN3261
RHKSOPCZBRMBHR-UHFFFAOYSA-N
bis(trifluoromethylsulfonyl)azanide;iron(3+)
134159302
Solid
Brown to Cream

Chemical Identifiers

207861-59-4
896.251
RHKSOPCZBRMBHR-UHFFFAOYSA-N
134159302
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Fe+3]
C6F18FeN3O12S6
MFCD23380171
Iron tris(trifluoromethanesulfonyl)imide; Iron tris(trifluoromethanesulfonimide)
bis(trifluoromethylsulfonyl)azanide;iron(3+)
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

SDS Documents