missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iron sulfide, 99.9% (metals basis), Thermo Scientific Chemicals
$83.15 - $475.63
Chemical Identifiers
CAS | 12068-85-8 |
---|---|
Molecular Formula | FeS2 |
Molecular Weight (g/mol) | 119.97 |
MDL Number | MFCD00064690 |
InChI Key | NFMAZVUSKIJEIH-UHFFFAOYSA-N |
Synonym | iron disulfide, iron sulfide, pyrite, bis sulfanylidene iron, disulfanylideneiron, iron disulfide iron pyrite, marcasite, naturally occuring mineral, grains, approximately 0.06-0.19 in, pyrite, naturally occurring mineral, grains, approximately 0.06-0.19in |
PubChem CID | 14788 |
SMILES | S=[Fe]=S |
Description
Iron sulfide is used in photovoltaics and lithium ion battery cathodes. It is also studied as a potential material for the sequestration of environmental contaminants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12068-85-8 | |
119.97 | |
NFMAZVUSKIJEIH-UHFFFAOYSA-N | |
14788 |
FeS2 | |
MFCD00064690 | |
iron disulfide, iron sulfide, pyrite, bis sulfanylidene iron, disulfanylideneiron, iron disulfide iron pyrite, marcasite, naturally occuring mineral, grains, approximately 0.06-0.19 in, pyrite, naturally occurring mineral, grains, approximately 0.06-0.19in | |
S=[Fe]=S |
Specifications
1171°C | |
12068-85-8 | |
1 g | |
FeS2 | |
iron disulfide, iron sulfide, pyrite, bis sulfanylidene iron, disulfanylideneiron, iron disulfide iron pyrite, marcasite, naturally occuring mineral, grains, approximately 0.06-0.19 in, pyrite, naturally occurring mineral, grains, approximately 0.06-0.19in | |
NFMAZVUSKIJEIH-UHFFFAOYSA-N | |
bis(sulfanylidene)iron | |
14788 | |
Odorless | |
4.87 |
Gray | |
Powder | |
99.99% (Metal basis) | |
MFCD00064690 | |
Insoluble in water. | |
S=[Fe]=S | |
119.97 | |
119.98 | |
Gray powder | |
Iron sulfide |
Safety and Handling
EINECSNumber : 235-106-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only