Please login to your online account to display your discounted pricing

Iron standard solution, for AAS, 1 mg/ml Fe in 2% HNO3, ACROS Organics™


Quantity 100mL
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  196051000

 View more versions of this product

Catalog No. AC196051000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 55.85
Quantity 100mL
Color Clear colorless
Purity Grade For AAS
Concentration 997 to 1003 μg/mL of Fe (3+) in 2% HNO3 (20°C)
Density 7.86
CAS 7697-37-2
CAS 7732-18-5
Health Hazard 1 GHS Signal Word: Warning
Name Note 1 mg/ml Fe in 2% HNO3
MDL Number MFCD00010999
CAS 7439-89-6
Chemical Name or Material Iron standard solution
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
Keep only in original container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Plastic bottle
Molecular Formula Fe
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May be corrosive to metals.

Specifications

Chemical Name or Material Iron standard solution
Name Note 1 mg/ml Fe in 2% HNO3
CAS 7439-89-6
CAS 7697-37-2
CAS 7732-18-5
Purity Grade For AAS
Molecular Formula Fe
Formula Weight 55.85
MDL Number MFCD00010999
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face, hands and any exposed skin thoroughly after handling.
Keep only in original container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May be corrosive to metals.

SDS & Safety Documents