Please login to your online account to display your discounted pricing

Iron Standard Solution, 50ppm (50μg/mL), LabChem™

$97.11 - $97.11

Chemical Identifiers

Chemical Name or Material Iron Standard Solution
CAS 7732-18-5
CAS Max % 99.91
CAS 7664-93-9
CAS Max % 0.05
CAS 7783-85-9
CAS Max % 0.04
Name Note 50ppm (50μg/mL)
Linear Formula Fe(NH4)2(SO4)2·6H2O
Formula Weight 392.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC157301

 
lab chem
LC157301
Poly Bottle 500mL Each for $97.11
Description & Specifications

Specifications

Formula Weight 392.14
Name Note 50ppm (50μg/mL)
CAS Max % 99.91
Solubility Soluble in water
Quantity 500mL
Density 1g/mL
Color Clear, colorless
Recommended Storage Room Temperature
CAS Max % 0.04
CAS 7783-85-9
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS 7732-18-5
Chemical Name or Material Iron Standard Solution
Concentration 50ppm (50μg/mL)
CAS Max % 0.05
Packaging Poly Bottle
CAS 7664-93-9
Physical Form Liquid
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Linear Formula Fe(NH4)2(SO4)2·6H2O
Identification Passes Test
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature