missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis)

$788.11 - $788.11

Chemical Identifiers

CAS 7439-89-6
MDL Number MFCD00010999
Chemical Name or Material Iron sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA40857KS SDS Alfa Aesar™
40857KS
1 each Each for $788.11
Specifications Description & Specifications

Chemical Identifiers

7439-89-6
MFCD00010999
Iron sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick

Specifications

1 each
MFCD00010999
99.95%, (metals basis)
7439-89-6
Iron sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick

Safety and Handling

TSCA : TSCA