Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis)

$590.65 - $590.65

Chemical Identifiers

Chemical Name or Material Iron sputtering target
CAS 7439-89-6
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00010999
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40855KS

  SDS
alfa aesar™
40855KS
1 each Each for $590.65
Description & Specifications

Specifications

Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
CAS 7439-89-6
Chemical Name or Material Iron sputtering target
TSCA TSCA
MDL Number MFCD00010999
Assay 99.95%
Quantity 1 each
Assay Percent Notes (metals basis)