Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis)

Magnetron sputtering source

$247.82 - $247.82

Chemical Identifiers

Chemical Name or Material Iron sputtering target
CAS 7439-89-6
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00010999
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40854KS

 
alfa aesar™
40854KS
1 each Each for $247.82
Description & Specifications

Specifications

Chemical Name or Material Iron sputtering target
TSCA TSCA
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.95%
Assay Percent Notes (metals basis)
MDL Number MFCD00010999
CAS 7439-89-6
Quantity 1 each