missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron rod, 12mm (0.47 in.) dia., Puratronic™, 99.995% (metals basis)

Manufacturer:  Alfa Aesar™ 11500HF

 View more versions of this product

Catalog No. AA11500HF



Specifications

Specifications

Iron rod
10mm
7439-89-6
Fe
3zhp
[Fe]
55.845
CHEBI:82664
99.995%
≈0.89g/mm
12mm (0.47 in.) dia.
(metals basis)
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
23925
Rod
Puratronic™

Chemical Identifiers

7439-89-6
55.845
XEEYBQQBJWHFJM-UHFFFAOYSA-N
23925
iron
Fe
MFCD00010999
3zhp
CHEBI:82664
[Fe]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents