Please login to your online account to display your discounted pricing

Iron, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

For atomic absorption. (1mL = 1mg Fe) Solute: Ferric Nitrate Solvent: Dilute Nitric Acid

$78.00 - $121.18

Chemical Identifiers

Chemical Name or Material Iron, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 7782-61-8
CAS 7697-37-2
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00010999
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SI124-500

  SDS
fisher scientific company
SI124500
Poly Bottle 500mL Each for $121.18
Case of 6 Each for $727.08

SI124-100

  SDS
fisher scientific company
FLSI124100
Poly Bottle 100mL Each for $78.00
Case of 6 Each for $468.00
Description & Specifications

Specifications

Purity Grade Certified Grade
Melting Point 0°C
Boiling Point 100°C
Quantity 500mL
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
pH 1
Color Clear
Vapor Pressure 14mmHg
MDL Number MFCD00010999
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Concentration 990 to 1010 ppm
Traceability to NIST LOT 3126a Iron reference std.
Packaging Poly Bottle
CAS 7732-18-5
ChemAlert Storage Symbol White
CAS 7697-37-2
Name Note 1000ppm ±1%
Chemical Name or Material Iron, Reference Standard Solution
CAS 7782-61-8
Safety and Handling