missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iron powder, spherical, <20 micron, 99.5% (metals basis), Thermo Scientific Chemicals
$100.66 - $291.37
Chemical Identifiers
CAS | 7439-89-6 |
---|---|
Molecular Formula | Fe |
Molecular Weight (g/mol) | 55.85 |
MDL Number | MFCD00010999 |
InChI Key | XEEYBQQBJWHFJM-UHFFFAOYSA-N |
Synonym | ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m |
PubChem CID | 23925 |
ChEBI | CHEBI:82664 |
IUPAC Name | iron |
SMILES | [Fe] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-89-6 | |
55.85 | |
XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
23925 | |
iron |
Fe | |
MFCD00010999 | |
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m | |
CHEBI:82664 | |
[Fe] |
Specifications
7439-89-6 | |
>99.99% (metal basis), 99.5% | |
MFCD00010999 | |
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m | |
[Fe] | |
55.85 | |
CHEBI:82664 | |
Iron |
7.87g/cm3 at 20°C | |
Fe | |
25 g | |
XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
iron | |
23925 | |
Spherical Powder |
Safety and Handling
P235+P410-P280
H500-X
DOTInformation : DOT Class: 4.2; Packaging Group: II
EINECSNumber : 231-096-4
RTECSNumber : NO4565500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only