missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron Powder, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 7439-89-6
Physical Form Powder
Molecular Formula Fe
Formula Weight 55.85
Chemical Name or Material Iron, 100 Mesh
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK5312500 Avantor Performance
531212
Glass 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK5312212 Avantor Performance
531206
Glass 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7439-89-6
Fe
Iron, 100 Mesh
Powder
55.85

Specifications

55.85
Fe
Glass
Iron, 100 Mesh
500g
Powder
7439-89-6