missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron Powder, Macron Fine Chemicals™

macron

Manufacturer:  Avantor Performance 531212

 View more versions of this product

Catalog No. MK5312500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Iron
100 Mesh
Fe
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
55.845
CHEBI:82664
Powder
500g
7439-89-6
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
[Fe]
Glass
23925
55.85

Chemical Identifiers

7439-89-6
55.845
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
CHEBI:82664
[Fe]
Fe
XEEYBQQBJWHFJM-UHFFFAOYSA-N
23925
iron
Documents