Please login to your online account to display your discounted pricing

Iron Powder, Macron Fine Chemicals™


macron

Quantity 2.5kg
Packaging Glass

Manufacturer: avantor performance  531206

 View more versions of this product

Catalog No. MK5312212

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 55.85
Quantity 2.5kg
Chemical Name or Material Iron
Name Note 100 Mesh
Physical Form Powder
Molecular Formula Fe
CAS 7439-89-6
Packaging Glass

Specifications

Chemical Name or Material Iron
Name Note 100 Mesh
CAS 7439-89-6
Physical Form Powder
Molecular Formula Fe
Formula Weight 55.85