Please login to your online account to display your discounted pricing

Iron powder, 99.99% (metals basis), Alfa Aesar™, Puratronic™

$60.49 - $663.43

Chemical Identifiers

Chemical Name or Material Iron
CAS 7439-89-6
Physical Form Powder
Assay Percent Range 99.998% (Metals basis)
Molecular Formula Fe
MDL Number MFCD00010999
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1062106

 
alfa aesar
1062106
5g Each for $60.49

AA1062114

 
alfa aesar
1062114
25g Each for $226.27

AA1062122

 
alfa aesar
1062122
100g Each for $663.43
Description & Specifications

Specifications

Quantity 5g
Color Gray
DOT Information DOT Class: 4.1, Packaging Group: III
EINECS Number 231-096-4
Physical Form Powder
Chemical Name or Material Iron
Assay Percent Range 99.998% (Metals basis)
Molecular Formula Fe
MDL Number MFCD00010999
CAS 7439-89-6
Health Hazard 1 H228-H319-H335: Flammable solid. Causes serious eye irritation. May cause respiratory irritation.
Appearance Gray powder
Safety and Handling

H228-H319-H335: Flammable solid. Causes serious eye irritation. May cause respiratory irritation.