Alfa Aesar™ Iron, plasma standard solution, Specpure™, Fe 10,000μg/mL
Manufacturer:  Alfa Aesar™ 47221AD
 View more versions of this product
Catalog No. AA47221AD
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Iron, plasma standard solution
MFCD00010999
Liquid
Acidic
Fe 10,000μg/mL
UN3265
Specpure™
50mL
Chemical Identifiers
MFCD00010999
Safety and Handling
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : TSCA
SDS Documents