missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron pieces, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4509022

 View more versions of this product

Catalog No. AA4509022



Specifications

Specifications

Iron
7439-89-6
Fe
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
[Fe]
55.845
CHEBI:82664
100g
99.99%, metal basis
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
23925
Pieces, Irregularly Shaped, 3.2 To 6.4mm (0.125 To 0.25 in.)

Chemical Identifiers

7439-89-6
55.845
XEEYBQQBJWHFJM-UHFFFAOYSA-N
23925
iron
Fe
MFCD00010999
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
CHEBI:82664
[Fe]
Safety and Handling

Safety and Handling

EINECSNumber : 231-096-4

SDS Documents