Please login to your online account to display your discounted pricing

Iron phthalocyanine, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC412611000

 View more versions of this product

Catalog No. AC412611000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 568.38
Quantity 100g
Color Dark purple, green or black
Assay 70 to 90% (typically)
CAS 132-16-1
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material Iron phthalocyanine
Molecular Formula C32H16FeN8

Specifications

Chemical Name or Material Iron phthalocyanine
Assay 70 to 90% (typically)
Molecular Formula C32H16FeN8
Formula Weight 568.38