Please login to your online account to display your discounted pricing

Iron pentacarbonyl, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  412600250

 View more versions of this product

Catalog No. AC412600250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid Which May Contain Particulate Matter
Formula Weight 195.9
Quantity 25g
Color Yellow-orange to brown
Merck Index 15, 5143
TSCA TSCA
EINECS Number 236-670-8
Density 1.4900g/mL
RTECS Number NO4900000
Fieser 01,519; 02,229; 03,167; 04,268; 05,357; 06,304; 10,221; 13,152; 17,270
Packaging Glass bottle
Additional Information Material may have to be filtered through glass wool before use. Keep residue moist, may be pyrophoric if allowed to dry.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Synonym Pentacarbonyl iron
Health Hazard 2 GHS H Statement:
Fatal in contact with skin.
Fatal if inhaled.
Highly flammable liquid and vapour.
Fatal if swallowed.
Health Hazard 1 Danger
Chemical Name or Material Iron pentacarbonyl
CAS 13463-40-6
Name Note 99.5%
Assay Percent Range 99.50%
Linear Formula Fe(CO)5
Molecular Formula C5FeO5
MDL Number MFCD00011001

Specifications

Chemical Name or Material Iron pentacarbonyl
Name Note 99.5%
CAS 13463-40-6
Assay Percent Range 99.50%
Molecular Formula C5FeO5
Linear Formula Fe(CO)5
Formula Weight 195.9
MDL Number MFCD00011001
Synonym Pentacarbonyl iron
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Fatal in contact with skin.
Fatal if inhaled.
Highly flammable liquid and vapour.
Fatal if swallowed.

SDS & Safety Documents