missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Iron nitride, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 08819814
Description
Iron nitride is used in the synthesis of ammonia and carbon nanofibers. It is involved in the creation of ferrofluids, a liquid rocket fuel. It finds applications in making the strongest naturally magnetic material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Iron nitride | |
37245-77-5 | |
25 g | |
MFCD00134717 | |
Insoluble in water. | |
[NH2-].[N-]=[Fe].[Fe].[Fe].[Fe].[Fe].[Fe] | |
365.1 | |
Odorless |
Gray | |
Powder | |
Fe6H2N2-2 | |
iron nitride,-325 mesh powder, 1-aza-2,3,4-triferracyclobutan-1-yliron; diferrioamine | |
ILMFDGFXPJCFQW-UHFFFAOYSA-N | |
azanide;azanidylideneiron;iron | |
56843031 | |
Gray powder |
Chemical Identifiers
37245-77-5 | |
365.1 | |
ILMFDGFXPJCFQW-UHFFFAOYSA-N | |
56843031 | |
[NH2-].[N-]=[Fe].[Fe].[Fe].[Fe].[Fe].[Fe] |
Fe6H2N2-2 | |
MFCD00134717 | |
iron nitride,-325 mesh powder, 1-aza-2,3,4-triferracyclobutan-1-yliron; diferrioamine | |
azanide;azanidylideneiron;iron |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only