Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron(III) sulfate, Puratronic™, 99.998% (metals basis)

Chemical Identifiers

Chemical Name or Material Iron(III) sulfate
CAS 10028-22-5
Purity Grade Puratronic™
Formula Weight 399.87
MDL Number MFCD00011007
Merck Index 14,4032
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1071714

  SDS
alfa aesar™
1071714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1071706

  SDS
alfa aesar™
1071706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 399.87
Health Hazard 1 Warning
Purity Grade Puratronic™
TSCA TSCA
Quantity 25g
Odor Odorless
Assay Percent Notes (metals basis)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Solubility Information Slightly soluble in water
Sensitivity Hygroscopic
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Physical Form Powder
Chemical Name or Material Iron(III) sulfate
MDL Number MFCD00011007
Density 3.097g/mL
Merck Index 14,4032
CAS 10028-22-5
Assay 99.998%
Melting Point 480°C (dec.)
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.