missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(III) sulfate hydrate, Reagent Grade, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 033316A7

 View more versions of this product

Catalog No. AA33316A7



Description

Description

Iron(III) sulfate is also used in pigments, pickling baths for aluminum and steel. It acts as a mordant in dyeing, as a coagulant for industrial wastes. In the medical field, it is used as an astringent and styptic. It is involved in the preparation of aromatic esters from the corresponding acids and alcohols. Further, it is employed in the oxidation of pyrite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Iron(III) sulfate hydrate
Gray-White
5 kg
MFCD00011007
VXWSFRMTBJZULV-UHFFFAOYSA-H
iron(3+);trisulfate;hydrate
167265
Reagent
Hygroscopic
2.0-2.1
15244-10-7
Granules, Powder
Fe2H2O13S3
Soluble in waterSoluble in water. Sparingly soluble in alcohol.
O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
417.87
399.87 (Anhydrous)
Odorless
Grayish-white powder or granules

Chemical Identifiers

15244-10-7
417.87
VXWSFRMTBJZULV-UHFFFAOYSA-H
O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Fe2H2O13S3
MFCD00011007
167265
Safety and Handling

Safety and Handling

P201-P202-P264b-P270-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P310-P330-P332+P313-P362-P501c

H302-H315-H318-H350

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 233-072-9

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only