missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Iron(III) oxide, nanopowder, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 4411930

 View more versions of this product

Catalog No. AA4411930



Specifications

Specifications

Iron (III) oxide
(metals basis)
MFCD00011008
159.69
99.95%
Odorless
250m2/g
1565°C
1309-37-1
Fe2O3
14,4028
Nanopowder
5.24g/mL
3nm APS
Red to Brown
250g

Chemical Identifiers

1309-37-1
Fe2O3
MFCD00011008
Safety and Handling

Safety and Handling

RTECSNumber : NO7400000

TSCA : TSCA

Documents