missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(III) oxide, cosmetic, NanoArc™, Thermo Scientific Chemicals

Nanoarc Cosmetic Iron Oxide Is An Auburn Material offering Unique Cosmetic Qualitites. The Small Particle Size Results in a Strong Color Development Combined With High Translucency, For a Unique Glowing Look

Supplier:  Thermo Scientific Chemicals 04489536

 View more versions of this product

Catalog No. AA4489536



Description

Description

NanoArc cosmetic iron oxide is an auburn material offering unique cosmetic qualities. The small particle size results in a strong color development combined with high translucency for a unique glowing look.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

S.A. 30-60m2/g
Specifications

Specifications

Iron(III) oxide, cosmetic
Sold in collaboration with Nanophase Technologies Corporation
1309-37-1
500 g
MFCD00011008
JEIPFZHSYJVQDO-UHFFFAOYSA-N
trioxodiiron
159.69
Red-brown powder
1565°C
Red to Brown
Powder
Fe2O3
Insoluble in water
[Fe][Fe](=O)(=O)=O
159.69
Odorless
5.24

Chemical Identifiers

1309-37-1
159.69
JEIPFZHSYJVQDO-UHFFFAOYSA-N
[Fe][Fe](=O)(=O)=O
Fe2O3
MFCD00011008
trioxodiiron
Safety and Handling

Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 215-168-2

RTECSNumber : NO7400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only