missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Iron(III) oxide, cosmetic, Alfa Aesar™, NanoArc™

Nanoarc Cosmetic Iron Oxide Is An Auburn Material offering Unique Cosmetic Qualitites. The Small Particle Size Results in a Strong Color Development Combined With High Translucency, For a Unique Glowing Look

Manufacturer:  Alfa Aesar 4489522

 View more versions of this product

Catalog No. AA4489522



Description

Description

S.A. 30-60m2/g
Specifications

Specifications

Iron(III) oxide, cosmetic
1309-37-1
MFCD00011008
159.69
5.24g/cm3 at 20°C
Red to Brown
Sold in collaboration with Nanophase Technologies Corporation
Fe2O3
Red-brown powder
Powder
Odorless
100g

Chemical Identifiers

1309-37-1
Fe2O3
MFCD00011008
Safety and Handling

Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 215-168-2

SDS Documents